Assay Details
| Gene Symbol: LOC4339133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784345.2 | XP_015639831.1 | 4 - 5 | 531 | 60 | |
| GenBank mRNA | AK065541.1 | - | 4 - 5 | 535 | 60 | |
More Information
Set Membership: |
|

LOC4339133
Genomic Map