Assay Details
| Gene Symbol: LOC4339133 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784345.2 | XP_015639831.1 | 7 - 8 | 1356 | 70 | |
| GenBank mRNA | AK065541.1 | - | 7 - 8 | 1360 | 70 | |
| BT130050.1 | - | 1 - 2 | 106 | 70 | ||
More Information
Set Membership: |
|

LOC4339133
Genomic Map