Assay Details
| Gene Symbol: LOC4339190 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015783827.2 | XP_015639313.1 | 2 - 3 | 588 | 73 | |
| GenBank mRNA | AK059563.1 | - | 2 - 3 | 456 | 73 | |
| AK061025.1 | - | 1 - 2 | 885 | 73 | ||
More Information
Set Membership: |
|

LOC4339190
Genomic Map