Assay Details
| Gene Symbol: LOC4339242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782821.2 | XP_015638307.1 | 1 - 2 | 94 | 108 | |
| GenBank mRNA | AK064440.1 | - | 1 - 2 | 94 | 108 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015782821.2 transcripts.
Set Membership: |
|

LOC4339242
Genomic Map