Assay Details
| Gene Symbol: LOC4339242 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015782820.2 | XP_015638306.1 | 2 - 3 | 796 | 76 | |
| XM_015782821.2 | XP_015638307.1 | 2 - 3 | 802 | 76 | ||
| GenBank mRNA | AK064440.1 | - | 2 - 3 | 802 | 76 | |
More Information
Set Membership: |
|

LOC4339242
Genomic Map