Assay Details
| Gene Symbol: LOC4339322 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015784253.2 | XP_015639739.1 | 3 - 4 | 342 | 72 | |
| GenBank mRNA | AK111896.1 | - | 3 - 4 | 314 | 72 | |
| AK112031.1 | - | 3 - 4 | 313 | 72 | ||
More Information
Set Membership: |
|

LOC4339322
Genomic Map