Assay Details
| Gene Symbol: LOC4340001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785765.2 | XP_015641251.1 | 13 - 14 | 1465 | 123 | |
| GenBank mRNA | AK101132.1 | - | 13 - 14 | 1472 | 123 | |
More Information
Set Membership: |
|

LOC4340001
Genomic Map