Assay Details
| Gene Symbol: LOC4340001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785765.2 | XP_015641251.1 | 8 - 9 | 1137 | 73 | |
| XM_015785766.2 | XP_015641252.1 | 8 - 9 | 1130 | 73 | ||
| GenBank mRNA | AK101132.1 | - | 8 - 9 | 1144 | 73 | |
More Information
Set Membership: |
|

LOC4340001
Genomic Map