Assay Details
| Gene Symbol: LOC4340001 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015785765.2 | XP_015641251.1 | 9 - 10 | 1216 | 108 | |
| XM_015785766.2 | XP_015641252.1 | 9 - 10 | 1209 | 108 | ||
| GenBank mRNA | AK101132.1 | - | 9 - 10 | 1223 | 108 | |
More Information
Set Membership: |
|

LOC4340001
Genomic Map