Assay Details
| Gene Symbol: LOC9270139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789059.2 | XP_015644545.1 | 13 - 14 | 2142 | 69 | |
| GenBank mRNA | AK242714.1 | - | 1 - 2 | 80 | 69 | |
More Information
Set Membership: |
|

LOC9270139
Genomic Map