Assay Details
| Gene Symbol: LOC9270139 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789059.2 | XP_015644545.1 | 16 - 17 | 2487 | 56 | |
| GenBank mRNA | AK242714.1 | - | 4 - 5 | 425 | 56 | |
More Information
Set Membership: |
|

LOC9270139
Genomic Map