Assay Details
| Gene Symbol: LOC4340349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786098.2 | XP_015641584.1 | 15 - 16 | 2472 | 83 | |
| GenBank mRNA | AB037135.2 | - | 15 - 16 | 2473 | 83 | |
| AK103068.1 | - | 15 - 16 | 2426 | 83 | ||
More Information
Set Membership: |
|

LOC4340349
Genomic Map