Assay Details
| Gene Symbol: LOC4340349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786098.2 | XP_015641584.1 | 6 - 7 | 1509 | 72 | |
| XM_015786099.2 | XP_015641585.1 | 6 - 7 | 1461 | 72 | ||
| GenBank mRNA | AB037135.2 | - | 6 - 7 | 1510 | 72 | |
| AK103068.1 | - | 6 - 7 | 1463 | 72 | ||
More Information
Set Membership: |
|

LOC4340349
Genomic Map