Assay Details
| Gene Symbol: LOC4340349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786098.2 | XP_015641584.1 | 7 - 8 | 1625 | 125 | |
| XM_015786099.2 | XP_015641585.1 | 7 - 8 | 1577 | 125 | ||
| GenBank mRNA | AB037135.2 | - | 7 - 8 | 1626 | 125 | |
| AK103068.1 | - | 7 - 8 | 1579 | 125 | ||
More Information
Set Membership: |
|

LOC4340349
Genomic Map