Assay Details
| Gene Symbol: LOC4340495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786192.2 | XP_015641678.1 | 2 - 3 | 582 | 83 | |
| XM_015786193.2 | XP_015641679.1 | 2 - 3 | 572 | 83 | ||
| GenBank mRNA | AK111859.1 | - | 2 - 3 | 393 | 83 | |
More Information
Set Membership: |
|

LOC4340495
Genomic Map