Assay Details
| Gene Symbol: LOC4340495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786192.2 | XP_015641678.1 | 3 - 4 | 640 | 68 | |
| XM_015786193.2 | XP_015641679.1 | 3 - 4 | 630 | 68 | ||
| GenBank mRNA | AK111859.1 | - | 3 - 4 | 451 | 68 | |
| AY345223.2 | - | 3 - 4 | 392 | 68 | ||
More Information
Set Membership: |
|

LOC4340495
Genomic Map