Assay Details
Gene Symbol: LOC4340495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786192.2 | XP_015641678.1 | 4 - 5 | 742 | 57 | |
XM_015786193.2 | XP_015641679.1 | 4 - 5 | 732 | 57 | ||
GenBank mRNA | AY345223.2 | - | 4 - 5 | 494 | 57 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
