Assay Details
| Gene Symbol: LOC4340495 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786192.2 | XP_015641678.1 | 5 - 6 | 785 | 77 | |
| XM_015786193.2 | XP_015641679.1 | 5 - 6 | 775 | 77 | ||
| GenBank mRNA | AK111859.1 | - | 5 - 6 | 596 | 77 | |
| AY345223.2 | - | 5 - 6 | 537 | 77 | ||
More Information
Set Membership: |
|

LOC4340495
Genomic Map