Assay Details
Gene Symbol: LOC4340495 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015786192.2 | XP_015641678.1 | 7 - 8 | 975 | 94 | |
XM_015786193.2 | XP_015641679.1 | 7 - 8 | 965 | 94 | ||
GenBank mRNA | AK111859.1 | - | 7 - 8 | 786 | 94 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
