Assay Details
| Gene Symbol: LOC4340522 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788765.2 | XP_015644251.1 | 4 - 5 | 791 | 103 | |
| GenBank mRNA | AK072559.1 | - | 4 - 5 | 761 | 103 | |
More Information
Set Membership: |
|

LOC4340522
Genomic Map