Assay Details
| Gene Symbol: LOC4340529 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786482.2 | XP_015641968.2 | 5 - 6 | 427 | 61 | |
| GenBank mRNA | AK111776.1 | - | 5 - 6 | 434 | 61 | |
| FN663130.1 | - | 5 - 6 | 389 | 61 | ||
More Information
Set Membership: |
|

LOC4340529
Genomic Map