Assay Details
| Gene Symbol: LOC4341465 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789010.2 | XP_015644496.1 | 1 - 2 | 490 | 129 | |
| GenBank mRNA | AK063544.1 | - | 1 - 2 | 354 | 129 | |
| AK072067.1 | - | 1 - 2 | 467 | 129 | ||
More Information
Set Membership: |
|

LOC4341465
Genomic Map