Assay Details
| Gene Symbol: LOC4341774 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015786616.2 | XP_015642102.1 | 4 - 5 | 745 | 64 | |
| GenBank mRNA | AK119588.1 | - | 4 - 5 | 398 | 64 | |
More Information
Set Membership: |
|

LOC4341774
Genomic Map