Assay Details
| Gene Symbol: LOC4341897 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015788118.2 | XP_015643604.1 | 13 - 14 | 1483 | 108 | |
| XM_015788119.2 | XP_015643605.1 | 13 - 14 | 1430 | 108 | ||
| XM_015788121.2 | XP_015643607.1 | 14 - 15 | 1477 | 108 | ||
| GenBank mRNA | AK073794.1 | - | 4 - 5 | 1220 | 108 | |
| AK101144.1 | - | 13 - 14 | 1424 | 108 | ||
More Information
Set Membership: |
|

LOC4341897
Genomic Map