Assay Details
| Gene Symbol: LOC4342307 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792507.2 | XP_015647993.1 | 1 - 2 | 371 | 97 | |
| XM_015792508.2 | XP_015647994.1 | 1 - 2 | 371 | 97 | ||
| GenBank mRNA | AK073437.1 | - | 1 - 2 | 350 | 97 | |
 Back To Top
 Back To TopMore Information
| Set Membership: |  Amplicon length between 86 and 100  3' Most  Recommended Assay  Oryza sativa  Probe spans exons | 
Panther Classification:
 Back To Top
 Back To Top
 LOC4342307
LOC4342307 Genomic Map
Genomic Map Back To Top
 Back To Top