Assay Details
| Gene Symbol: LOC4342381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791970.2 | XP_015647456.1 | 3 - 4 | 438 | 87 | |
| GenBank mRNA | AK060737.1 | - | 3 - 4 | 431 | 87 | |
More Information
Set Membership: |
|

LOC4342381
Genomic Map