Assay Details
| Gene Symbol: LOC4342381 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791970.2 | XP_015647456.1 | 7 - 8 | 860 | 85 | |
| GenBank mRNA | AK060737.1 | - | 7 - 8 | 853 | 85 | |
| AK064848.1 | - | 7 - 8 | 860 | 85 | ||
More Information
Set Membership: |
|

LOC4342381
Genomic Map