Assay Details
Gene Symbol: LOC4342410 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AK069726.1 | - | 1 - 2 | 452 | 73 | |
AK242681.1 | - | 2497 | 73 | |||
EU703802.1 | - | 1 - 2 | 172 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
