Assay Details
| Gene Symbol: LOC4342513 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791988.2 | XP_015647474.1 | 4 - 5 | 858 | 83 | |
| GenBank mRNA | AF272978.1 | - | 4 - 5 | 398 | 83 | |
| AK062132.1 | - | 4 - 5 | 773 | 83 | ||
| AK066475.1 | - | 5 - 6 | 474 | 83 | ||
More Information
Set Membership: |
|

LOC4342513
Genomic Map