Assay Details
| Gene Symbol: LOC4342538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791204.2 | XP_015646690.1 | 10 - 11 | 2491 | 67 | |
| GenBank mRNA | AK062003.1 | - | 7 - 8 | 1137 | 67 | |
| AK065897.1 | - | 10 - 11 | 2493 | 67 | ||
More Information
Set Membership: |
|

LOC4342538
Genomic Map