Assay Details
| Gene Symbol: LOC4342538 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791204.2 | XP_015646690.1 | 4 - 5 | 1520 | 74 | |
| GenBank mRNA | AK062003.1 | - | 1 - 2 | 166 | 74 | |
| AK065897.1 | - | 4 - 5 | 1522 | 74 | ||
More Information
Set Membership: |
|

LOC4342538
Genomic Map