Assay Details
| Gene Symbol: LOC4342596 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789724.2 | XP_015645210.1 | 1 - 2 | 153 | 61 | |
| GenBank mRNA | AK122165.1 | - | 1 - 2 | 128 | 61 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015789724.2 transcripts.
Set Membership: |
|

LOC4342596
Genomic Map