Assay Details
| Gene Symbol: LOC4342826 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791680.2 | XP_015647166.1 | 9 - 10 | 1073 | 82 | |
| XM_015791681.2 | XP_015647167.1 | 9 - 10 | 1073 | 82 | ||
| XM_015791682.2 | XP_015647168.1 | 7 - 8 | 969 | 82 | ||
| GenBank mRNA | AK065341.1 | - | 9 - 10 | 1066 | 82 | |
| AK099348.1 | - | 9 - 10 | 1039 | 82 | ||
| AK104328.1 | - | 9 - 10 | 1060 | 82 | ||
| AK104826.1 | - | 9 - 10 | 1038 | 82 | ||
| JN848805.1 | - | 9 - 10 | 989 | 82 | ||
More Information
Set Membership: |
|

LOC4342826
Genomic Map