Assay Details
| Gene Symbol: LOC4342872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792502.2 | XP_015647988.1 | 12 - 13 | 2761 | 60 | |
| XM_015792503.2 | XP_015647989.1 | 12 - 13 | 2756 | 60 | ||
| XM_015792504.2 | XP_015647990.1 | 9 - 10 | 1448 | 60 | ||
| GenBank mRNA | AK070203.1 | - | 11 - 12 | 2638 | 60 | |
More Information
Set Membership: |
|

LOC4342872
Genomic Map