Assay Details
| Gene Symbol: LOC4342872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792502.2 | XP_015647988.1 | 3 - 4 | 412 | 115 | |
| XM_015792503.2 | XP_015647989.1 | 3 - 4 | 407 | 115 | ||
| GenBank mRNA | AK070203.1 | - | 2 - 3 | 289 | 115 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_015792502.2, XM_015792503.2 transcripts.
Set Membership: |
|

LOC4342872
Genomic Map