Assay Details
| Gene Symbol: LOC4342872 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792502.2 | XP_015647988.1 | 5 - 6 | 1576 | 108 | |
| XM_015792503.2 | XP_015647989.1 | 5 - 6 | 1571 | 108 | ||
| XM_015792504.2 | XP_015647990.1 | 2 - 3 | 263 | 108 | ||
| GenBank mRNA | AK070203.1 | - | 4 - 5 | 1453 | 108 | |
More Information
Set Membership: |
|

LOC4342872
Genomic Map