Assay Details
| Gene Symbol: LOC4342961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789320.2 | XP_015644806.1 | 4 - 5 | 785 | 89 | |
| GenBank mRNA | AK060569.1 | - | 4 - 5 | 720 | 89 | |
| AK073103.1 | - | 4 - 5 | 754 | 89 | ||
More Information
Set Membership: |
|

LOC4342961
Genomic Map