Assay Details
| Gene Symbol: LOC4342961 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789320.2 | XP_015644806.1 | 5 - 6 | 852 | 65 | |
| GenBank mRNA | AK060569.1 | - | 5 - 6 | 787 | 65 | |
| AK073103.1 | - | 5 - 6 | 821 | 65 | ||
More Information
Set Membership: |
|

LOC4342961
Genomic Map