Assay Details
| Gene Symbol: LOC4343145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790169.2 | XP_015645655.1 | 2 - 3 | 841 | 128 | |
| XM_015790170.2 | XP_015645656.1 | 2 - 3 | 836 | 128 | ||
| XM_015790171.2 | XP_015645657.1 | 2 - 3 | 841 | 128 | ||
| GenBank mRNA | AK242842.1 | - | 2 - 3 | 820 | 128 | |
More Information
Set Membership: |
|

LOC4343145
Genomic Map