Assay Details
| Gene Symbol: LOC4343145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015790169.2 | XP_015645655.1 | 6 - 7 | 1383 | 132 | |
| XM_015790170.2 | XP_015645656.1 | 6 - 7 | 1378 | 132 | ||
| XM_015790171.2 | XP_015645657.1 | 6 - 7 | 1383 | 132 | ||
| GenBank mRNA | AK242842.1 | - | 5 - 6 | 1944 | 132 | |
More Information
Set Membership: |
|

LOC4343145
Genomic Map