Assay Details
| Gene Symbol: LOC9272191 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015789821.2 | XP_015645307.1 | 2 - 3 | 320 | 142 | |
| XM_015789822.2 | XP_015645308.1 | 2 - 3 | 323 | 142 | ||
| GenBank mRNA | AK073933.1 | - | 2 - 3 | 213 | 142 | |
More Information
Set Membership: |
|

LOC9272191
Genomic Map