Assay Details
| Gene Symbol: LOC4343216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791911.2 | XP_015647397.1 | 5 - 6 | 830 | 86 | |
| XM_015791912.2 | XP_015647398.1 | 5 - 6 | 829 | 86 | ||
| XM_015791913.2 | XP_015647399.1 | 5 - 6 | 549 | 86 | ||
| GenBank mRNA | AK066198.1 | - | 4 - 5 | 721 | 86 | |
More Information
Set Membership: |
|

LOC4343216
Genomic Map