Assay Details
| Gene Symbol: LOC4343216 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791911.2 | XP_015647397.1 | 10 - 11 | 1414 | 86 | |
| XM_015791912.2 | XP_015647398.1 | 10 - 11 | 1413 | 86 | ||
| XM_015791913.2 | XP_015647399.1 | 10 - 11 | 1133 | 86 | ||
| GenBank mRNA | AK066198.1 | - | 9 - 10 | 1305 | 86 | |
More Information
Set Membership: |
|

LOC4343216
Genomic Map