Assay Details
| Gene Symbol: LOC4343217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791881.2 | XP_015647367.1 | 15 - 16 | 2121 | 128 | |
| GenBank mRNA | AB057651.1 | - | 15 - 16 | 1957 | 128 | |
| AK067379.1 | - | 15 - 16 | 2106 | 128 | ||
| AK121737.1 | - | 15 - 16 | 2099 | 128 | ||
More Information
Set Membership: |
|

LOC4343217
Genomic Map