Assay Details
| Gene Symbol: LOC4343217 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791881.2 | XP_015647367.1 | 9 - 10 | 1434 | 116 | |
| GenBank mRNA | AB057651.1 | - | 9 - 10 | 1270 | 116 | |
| AK067379.1 | - | 9 - 10 | 1413 | 116 | ||
| AK121737.1 | - | 9 - 10 | 1412 | 116 | ||
More Information
Set Membership: |
|

LOC4343217
Genomic Map