Assay Details
| Gene Symbol: LOC9271882 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015792087.2 | XP_015647573.1 | 1 - 2 | 2659 | 69 | |
| GenBank mRNA | AK119387.1 | - | 1 - 2 | 2569 | 69 | |
More Information
Set Membership: |
|

LOC9271882
Genomic Map