Assay Details
| Gene Symbol: LOC4343565 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791010.2 | XP_015646496.1 | 11 - 12 | 5594 | 130 | |
| GenBank mRNA | AK058724.1 | - | 1 - 2 | 195 | 130 | |
| AK066706.1 | - | 3 - 4 | 2467 | 130 | ||
| AK100968.1 | - | 3 - 4 | 1718 | 130 | ||
More Information
Set Membership: |
|

LOC4343565
Genomic Map