Assay Details
| Gene Symbol: LOC4343678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791554.2 | XP_015647040.1 | 4 - 5 | 762 | 89 | |
| GenBank mRNA | AK060818.1 | - | 2 - 3 | 281 | 89 | |
| AK100426.1 | - | 4 - 5 | 657 | 89 | ||
More Information
Set Membership: |
|

LOC4343678
Genomic Map