Assay Details
| Gene Symbol: LOC4343678 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_015791554.2 | XP_015647040.1 | 6 - 7 | 1358 | 150 | |
| GenBank mRNA | AK060818.1 | - | 3 - 4 | 986 | 150 | |
| AK100426.1 | - | 6 - 7 | 1253 | 150 | ||
More Information
Set Membership: |
|

LOC4343678
Genomic Map