Assay Details
Gene Symbol: LOC4343808 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_015789113.2 | XP_015644599.1 | 6 - 7 | 1971 | 85 | |
GenBank mRNA | AK069667.1 | - | 6 - 7 | 1932 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
